siRNA Details
Gene Symbol: Vmn1r216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134245.1 | NP_599006.1 | 1 | 813 | ||
GenBank mRNA | BC145650.1 | - | 1 | 891 | 40130953 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
