siRNA Details
Gene Symbol: Acot3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134246.3 | NP_599007.1 | 4 | 1574 | ||
GenBank mRNA | AY563097.1 | - | 4 | 1577 | ||
AY563098.1 | - | 4 | 834 | |||
BC138741.1 | - | 3 | 1173 | 8861763 | ||
BC138743.1 | - | 3 | 1174 | 8861765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
