siRNA Details
Gene Symbol: Havcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF450241.1 | - | 1, 2 | 133 | ||
BC106851.2 | - | 1, 2 | 95 | 40046221 | ||
BC106852.2 | - | 1, 2 | 95 | 40046222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
