siRNA Details
Gene Symbol: Apobr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138310.1 | NP_612183.1 | 2 | 1140 | ||
GenBank mRNA | AF141335.1 | - | 2 | 1140 | ||
AK156422.1 | - | 2 | 1208 | |||
BC030718.1 | - | 2 | 1148 | 4482457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
