siRNA Details
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133930.1 | NP_598691.1 | 2 | 571 | ||
GenBank mRNA | AB009020.1 | - | 2 | 576 | ||
AK042344.1 | - | 2 | 718 | |||
AK077726.1 | - | 2 | 715 | |||
AK155777.1 | - | 2 | 581 | |||
BC010804.1 | - | 2 | 571 | 4239786 | ||
BC023893.1 | - | 2 | 582 | 5310111 | ||
BC025932.1 | - | 2 | 477 | 5102571 | ||
BC029065.1 | - | 2 | 607 | 4935296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
