siRNA Details
Gene Symbol: Helq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081107.1 | NP_001074576.1 | 9 | 1874 | ||
GenBank mRNA | AF436846.1 | - | 7 | 856 | ||
AK052427.1 | - | 9 | 1834 | |||
BC082601.1 | - | 9 | 1838 | 30662103 | ||
BC109169.2 | - | 9 | 1742 | |||
BC109170.2 | - | 9 | 1747 | 40048258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
