siRNA Details
Gene Symbol: Socs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138657.3 | NP_619598.1 | 3, 4 | 1044 | ||
GenBank mRNA | AF424814.1 | - | 3, 4 | 1045 | ||
AK147580.1 | - | 3, 4 | 1142 | |||
AK147650.1 | - | 3, 4 | 1142 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006247484.
Gene Ontology Categories:
