siRNA Details
Gene Symbol: Prpf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138659.2 | NP_619600.2 | 36 | 6194 | ||
GenBank mRNA | AB047391.1 | - | 36 | 5920 | ||
AK077412.1 | - | 5 | 568 | |||
AK167984.1 | - | 1 | 48 | |||
BC034648.1 | - | 16 | 2477 | |||
BC093481.1 | - | 36 | 6124 | 30945329 | ||
CO426130.1 | - | 4 | 445 |
