siRNA Details
Gene Symbol: Casc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138660.2 | NP_619601.2 | 9 | 1682 | ||
GenBank mRNA | AF526276.1 | - | 9 | 1656 | ||
AJ292072.1 | - | 1896 | ||||
AK037399.1 | - | 3 | 277 | |||
AK139608.1 | - | 9 | 1721 | |||
AK145886.1 | - | 9 | 1456 | |||
BC034533.1 | - | 6 | 1326 | |||
BC060672.1 | - | 9 | 1510 | |||
BC141296.1 | - | 9 | 1795 | 9055908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
