siRNA Details
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138669.1 | NP_619610.1 | 8 | 853 | ||
GenBank mRNA | AK075920.1 | - | 8 | 829 | ||
AK150277.1 | - | 8 | 869 | |||
AK151786.1 | - | 8 | 831 | |||
AK152824.1 | - | 8 | 869 | |||
AK153359.1 | - | 8 | 835 | |||
AK157855.1 | - | 8 | 829 | |||
AK167107.1 | - | 8 | 891 | |||
AK168319.1 | - | 8 | 833 | |||
AK169350.1 | - | 8 | 833 | |||
AK169815.1 | - | 2259 | ||||
BC008132.1 | - | 8 | 853 | 3498954 | ||
BC012862.1 | - | 8 | 841 | 3585696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
