siRNA Details
Gene Symbol: Flna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290421.1 | NP_001277350.1 | 45 | 7547 | ||
NM_010227.3 | NP_034357.2 | 44 | 7525 | |||
GenBank mRNA | AK131179.1 | - | 32 | 5366 | ||
BC038478.1 | - | 45 | 7422 | |||
BC039208.1 | - | 24 | 3737 | |||
BC054432.1 | - | 1 | 125 | |||
BC099506.1 | - | 2 | 297 | |||
BC151024.1 | - | 44 | 7516 | 9087937 |
