siRNA Details
Gene Symbol: Nadk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159637.1 | NP_001153109.1 | 3 | 402 | ||
NM_138671.2 | NP_619612.2 | 3 | 301 | |||
GenBank mRNA | AK147917.1 | - | 3 | 379 | ||
AK165510.1 | - | 3 | 395 | |||
AK170095.1 | - | 3 | 383 | |||
AK170608.1 | - | 3 | 397 | |||
AK170960.1 | - | 3 | 383 | |||
BC004012.1 | - | 3 | 289 | 3493623 | ||
CJ166231.1 | - | 3 | 402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
