siRNA Details
Gene Symbol: Stab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138673.2 | NP_619614.1 | 26 | 2920 | ||
GenBank mRNA | AB105393.1 | - | 26 | 2920 | ||
AB105396.1 | - | 26 | 2965 | |||
AB105397.1 | - | 26 | 2997 | |||
AB105398.1 | - | 26 | 2965 | |||
AB105399.1 | - | 26 | 2965 | |||
AF364951.2 | - | 26 | 2920 | |||
AK142339.1 | - | 5 | 489 | |||
BC145110.1 | - | 25 | 2743 | |||
DQ786775.1 | - | 26 | 2916 |
