siRNA Details
Gene Symbol: Tmem47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138751.2 | NP_620090.1 | 3 | 699 | ||
GenBank mRNA | AB041540.1 | - | 3 | 632 | ||
AK030761.1 | - | 3 | 628 | |||
AK076147.1 | - | 3 | 638 | |||
AK133790.1 | - | 3 | 636 | |||
AK162304.1 | - | 3 | 637 | |||
BC019751.1 | - | 3 | 610 | 4020610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
