siRNA Details
Gene Symbol: Tmlhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138758.1 | NP_620097.1 | 4, 5 | 759 | ||
GenBank mRNA | AK052425.1 | - | 4, 5 | 761 | ||
AK144627.1 | - | 4, 5 | 755 | |||
AK161724.1 | - | 4, 5 | 726 | |||
AY033513.1 | - | 3, 4 | 597 | |||
BC010495.1 | - | 4, 5 | 759 | 4195680 | ||
BC115365.1 | - | 4, 5 | 691 | 40059106 |
