siRNA Details
Gene Symbol: Ripk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138952.3 | NP_620402.1 | 4, 5 | 737 | ||
GenBank mRNA | AF461040.1 | - | 4, 5 | 635 | ||
AF487539.1 | - | 4, 5 | 635 | |||
AK159593.1 | - | 4, 5 | 761 | |||
AK172319.1 | - | 4, 5 | 759 | |||
AK172417.1 | - | 4, 5 | 759 | |||
BC025611.1 | - | 2, 3 | 255 | |||
BC050845.1 | - | 4, 5 | 750 | |||
BC058088.1 | - | 4, 5 | 882 | |||
BC069878.1 | - | 4, 5 | 762 | 5316066 |

More Information
Other Species Targeted

Gene Ontology Categories:
