siRNA Details
Gene Symbol: Arhgdia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133796.7 | NP_598557.3 | 3 | 333 | ||
GenBank mRNA | AK075656.1 | - | 2 | 200 | ||
AK077343.1 | - | 3 | 493 | |||
AK143516.1 | - | 3 | 317 | |||
AK147842.1 | - | 292 | ||||
AK156819.1 | - | 278 | ||||
AK165241.1 | - | 3 | 315 | |||
AK170553.1 | - | 3 | 300 | |||
AK170718.1 | - | 3 | 317 | |||
AK172140.1 | - | 278 | ||||
BC004732.1 | - | 3 | 275 | 3583026 | ||
BC086755.1 | - | 3 | 257 | 6414768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
