siRNA Details
Gene Symbol: Rassf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138956.3 | NP_620406.1 | 4 | 651 | ||
GenBank mRNA | AF332864.1 | - | 4 | 662 | ||
AK155495.1 | - | 4 | 678 | |||
AK167627.1 | - | 4 | 484 | |||
AK196922.1 | - | 1 | 28 | |||
BC011511.1 | - | 4 | 651 | 3495833 | ||
BC055023.1 | - | 4 | 706 | 5250629 | ||
BC071204.1 | - | 4 | 659 | 2648929 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
