siRNA Details
Gene Symbol: Lrrc75a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198861.1 | NP_942561.1 | 3 | 563 | ||
GenBank mRNA | AK146714.1 | - | 3 | 252 | ||
AK186775.1 | - | 1 | 19 | |||
BC046404.1 | - | 3 | 303 | |||
BC053455.1 | - | 3 | 563 | 6513708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
