siRNA Details
Gene Symbol: Rnf185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290472.1 | NP_001277401.1 | 4 | 1002 | ||
NM_001290473.1 | NP_001277402.1 | 3 | 983 | |||
NM_145355.5 | NP_663330.2 | 5 | 1121 | |||
GenBank mRNA | AK034108.1 | - | 5 | 491 | ||
AK052436.1 | - | 4 | 411 | |||
AK053358.1 | - | 4 | 368 | |||
AK075756.1 | - | 4 | 1003 | |||
AK077638.1 | - | 4 | 333 | |||
AK149640.1 | - | 4 | 354 | |||
AK165895.1 | - | 427 | ||||
AK167455.1 | - | 4 | 319 | |||
AK168904.1 | - | 4 | 356 | |||
BC014812.1 | - | 4 | 333 | 3153870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
