siRNA Details
Gene Symbol: Hspa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010479.2 | NP_034609.2 | 1 | 2758 | ||
GenBank mRNA | AK132331.1 | - | 1 | 2758 | ||
AK171820.1 | - | 2741 | ||||
BC054782.1 | - | 1 | 2726 | 6815894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
