siRNA Details
Gene Symbol: Mcfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139295.3 | NP_647456.1 | 3 | 389 | ||
NM_176808.5 | NP_789778.1 | 4 | 614 | |||
GenBank mRNA | AF475283.1 | - | 3 | 357 | ||
AK033139.1 | - | 3 | 323 | |||
AK075781.1 | - | 4 | 547 | |||
AK077139.1 | - | 3 | 340 | |||
AK077631.1 | - | 3 | 323 | |||
AK141599.1 | - | 4 | 572 | |||
AK151803.1 | - | 3 | 378 | |||
AK211007.1 | - | 1 | 70 | |||
BC003996.1 | - | 3 | 316 | 3492270 | ||
GQ896355.1 | - | 2 | 260 | |||
GQ896356.1 | - | 2 | 260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
