siRNA Details
Gene Symbol: Gm13023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007077.2 | NP_001007078.2 | 3 | 448 | ||
GenBank mRNA | AK135972.1 | - | 3 | 431 | ||
BC084594.1 | - | 3 | 429 | 30478700 | ||
CA559385.1 | - | 3 | 418 | |||
CF174133.1 | - | 3 | 456 |
