siRNA Details
Gene Symbol: Trpv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007572.2 | NP_001007573.1 | 15 | 1963 | ||
GenBank mRNA | AK085479.1 | - | 15 | 2010 | ||
BC110554.1 | - | 15 | 1963 | 40047811 | ||
BC110555.1 | - | 15 | 1963 | 40047816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
