siRNA Details
Gene Symbol: Obox7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038676.1 | NP_001033765.1 | 2 | 213 | ||
GenBank mRNA | BC099488.1 | - | 2 | 213 | 30939375 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Obox7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038676.1 | NP_001033765.1 | 2 | 213 | ||
GenBank mRNA | BC099488.1 | - | 2 | 213 | 30939375 |