siRNA Details
Gene Symbol: Btg1-ps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034856.2 | NP_001030028.1 | 5 | 734 | ||
GenBank mRNA | AK132831.1 | - | 5 | 736 | ||
BC147535.1 | - | 2 | 324 | 9056526 | ||
BC147539.1 | - | 2 | 324 | 9056530 |
Gene Symbol: Btg1-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243943.1 | NP_001230872.1 | 5 | 513 | ||
GenBank mRNA | AK132937.1 | - | 5 | 514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
