siRNA Details
Gene Symbol: Slc25a43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085497.2 | NP_001078966.1 | 4 | 921 | ||
GenBank mRNA | AK153028.1 | - | 3 | 602 | ||
BC147709.1 | - | 4 | 921 | 9087468 | ||
BC147717.1 | - | 4 | 921 | 9087476 | ||
BC172137.1 | - | 5 | 932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
