siRNA Details
Gene Symbol: Gm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033234.2 | NP_001028406.1 | 2, 3 | 534 | ||
GenBank mRNA | AK131663.1 | - | 2, 3 | 499 | ||
AK145837.1 | - | 2, 3 | 535 | |||
BC048624.1 | - | 2, 3 | 498 |
