siRNA Details
Gene Symbol: Sun3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290519.1 | NP_001277448.1 | 6 | 426 | ||
NM_001290520.1 | NP_001277449.1 | 4 | 349 | |||
NM_177576.3 | NP_808244.1 | 5 | 453 | |||
GenBank mRNA | AK052771.1 | - | 5 | 454 | ||
AK132922.1 | - | 6 | 427 | |||
BC109334.2 | - | 4 | 263 | 40051782 | ||
HQ141857.1 | - | 6 | 416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
