siRNA Details
Gene Symbol: Tmem199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199199.3 | NP_954669.2 | 2 | 287 | ||
GenBank mRNA | AK156856.1 | - | 2 | 222 | ||
AK158039.1 | - | 2 | 251 | |||
BC050977.1 | - | 2 | 250 | |||
BC147070.1 | - | 2 | 275 | 9056057 | ||
BC147071.1 | - | 2 | 275 | 9056065 | ||
BY255000.1 | - | 2 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
