siRNA Details
Gene Symbol: Nlrp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004142.2 | NP_001004142.2 | 6 | 2421 | ||
GenBank mRNA | AY355339.1 | - | 6 | 2362 | ||
DQ117601.1 | - | 5 | 2362 | |||
KC539856.1 | - | 5 | 2203 | |||
KC539857.1 | - | 5 | 2203 | |||
KC539858.1 | - | 5 | 2203 | |||
KC539859.1 | - | 5 | 2203 | |||
KC539860.1 | - | 5 | 2203 | |||
KC539861.1 | - | 5 | 2203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
