siRNA Details
Gene Symbol: Nlrp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004142.2 | NP_001004142.2 | 3 | 392 | ||
GenBank mRNA | AK157034.1 | - | 3 | 393 | ||
AY355339.1 | - | 2 | 174 | |||
DQ117601.1 | - | 2 | 333 | |||
KC539856.1 | - | 2 | 174 | |||
KC539857.1 | - | 2 | 174 | |||
KC539858.1 | - | 2 | 174 | |||
KC539859.1 | - | 2 | 174 | |||
KC539860.1 | - | 2 | 174 | |||
KC539861.1 | - | 2 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
