siRNA Details
Gene Symbol: Trim40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033235.3 | NP_001028407.1 | 2 | 780 | ||
NM_001172168.1 | NP_001165639.1 | 2 | 780 | |||
GenBank mRNA | AK136498.1 | - | 2 | 700 | ||
BC147336.1 | - | 1 | 471 | 9056401 | ||
BC147337.1 | - | 1 | 471 | 9056402 | ||
BC172037.1 | - | 1 | 471 | 9054323 |
