siRNA Details
Gene Symbol: Gm13152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039209.2 | NP_001034298.1 | 452 | |||
GenBank mRNA | BC058613.1 | - | 452 | 30044792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
