siRNA Details
Gene Symbol: Stxbp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114612.1 | NP_001108084.1 | 21 | 2482 | ||
NM_172440.3 | NP_766028.2 | 22 | 2554 | |||
GenBank mRNA | AK035159.1 | - | 21 | 2448 | ||
AY542324.1 | - | 21 | 2405 | |||
AY542326.1 | - | 20 | 2333 | |||
BC137883.1 | - | 21 | 2354 | 8860902 | ||
BC145008.1 | - | 22 | 2426 | 9053556 | ||
BC145009.1 | - | 22 | 2447 | 9053557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
