siRNA Details
Gene Symbol: Stxbp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114611.1 | NP_001108083.1 | 21 | 2378 | ||
NM_001114612.1 | NP_001108084.1 | 22 | 2549 | |||
NM_001114613.1 | NP_001108085.1 | 22 | 2450 | |||
NM_172440.3 | NP_766028.2 | 23 | 2621 | |||
GenBank mRNA | AK035159.1 | - | 22 | 2515 | ||
AY542324.1 | - | 22 | 2472 | |||
AY542325.1 | - | 20 | 2229 | |||
AY542326.1 | - | 21 | 2400 | |||
AY542327.1 | - | 21 | 2301 | |||
BC137883.1 | - | 22 | 2421 | 8860902 | ||
BC145007.1 | - | 23 | 2415 | 9053555 | ||
BC145008.1 | - | 23 | 2493 | 9053556 | ||
BC145009.1 | - | 23 | 2514 | 9053557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
