siRNA Details
Gene Symbol: Stxbp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114611.1 | NP_001108083.1 | 22 | 2920 | ||
NM_001114612.1 | NP_001108084.1 | 23 | 3091 | |||
NM_001114613.1 | NP_001108085.1 | 23 | 2992 | |||
NM_172440.3 | NP_766028.2 | 24 | 3163 | |||
GenBank mRNA | AK035159.1 | - | 23 | 3057 | ||
AY542324.1 | - | 23 | 3014 | |||
AY542325.1 | - | 21 | 2771 | |||
AY542326.1 | - | 22 | 2942 | |||
AY542327.1 | - | 22 | 2843 | |||
BC137883.1 | - | 23 | 2963 | 8860902 | ||
BC145007.1 | - | 24 | 2957 | 9053555 | ||
BC145008.1 | - | 24 | 3035 | 9053556 | ||
BC145009.1 | - | 24 | 3056 | 9053557 | ||
BU609516.1 | - | 1 | 234 |

More Information
Other Species Targeted

Gene Ontology Categories:
