siRNA Details
Gene Symbol: Elfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183141.2 | NP_898964.2 | 2 | 1714 | ||
GenBank mRNA | AK031970.1 | - | 1 | 1304 | ||
AK173294.1 | - | 1 | 180 | |||
BC076582.1 | - | 2 | 1955 | |||
BC079588.1 | - | 1 | 1214 | 5706857 | ||
BC094219.1 | - | 2 | 1714 | 6840886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
