siRNA Details
Gene Symbol: Camkk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199676.1 | NP_001186605.1 | 8, 9 | 1038 | ||
NM_145358.2 | NP_663333.1 | 8, 9 | 1038 | |||
GenBank mRNA | AF453383.1 | - | 7, 8 | 821 | ||
AK031399.1 | - | 8, 9 | 963 | |||
AK032070.1 | - | 8, 9 | 1011 | |||
AK044660.1 | - | 8, 9 | 1027 | |||
AK122370.1 | - | 3, 4 | 204 | |||
BC023103.1 | - | 7, 8 | 871 | 3993474 |
