siRNA Details
Gene Symbol: Camkk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199676.1 | NP_001186605.1 | 6 | 930 | ||
NM_145358.2 | NP_663333.1 | 6 | 930 | |||
GenBank mRNA | AF453383.1 | - | 5 | 713 | ||
AK031399.1 | - | 6 | 855 | |||
AK032070.1 | - | 6 | 903 | |||
AK044660.1 | - | 6 | 919 | |||
AK122370.1 | - | 1 | 96 | |||
BC023103.1 | - | 5 | 763 | 3993474 |
