siRNA Details
Gene Symbol: Wdr37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039388.2 | NP_001034477.1 | 2 | 348 | ||
NM_001039389.2 | NP_001034478.1 | 2 | 269 | |||
NM_172445.3 | NP_766033.1 | 2 | 269 | |||
GenBank mRNA | AK014128.1 | - | 2 | 245 | ||
AK032582.1 | - | 2 | 231 | |||
AK036203.1 | - | 2 | 265 | |||
AK044740.1 | - | 2 | 249 | |||
AK129254.1 | - | 2 | 225 | |||
AK146209.1 | - | 2 | 244 | |||
BB611667.1 | - | 2 | 348 | |||
BC046236.1 | - | 1 | 96 | 6817089 | ||
BC048819.1 | - | 2 | 193 | |||
CJ134699.1 | - | 2 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
