siRNA Details
Gene Symbol: Gtpbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153116.1 | NP_694756.1 | 8 | 802 | ||
GenBank mRNA | AK139595.1 | - | 8 | 816 | ||
AK160252.1 | - | 8 | 841 | |||
BC034507.1 | - | 8 | 828 | 4222393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
