siRNA Details
Gene Symbol: Csrnp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153407.2 | NP_700456.1 | 5 | 1608 | ||
GenBank mRNA | AB091689.1 | - | 4 | 1357 | ||
AK050864.1 | - | 5 | 1629 | |||
AK146924.1 | - | 5 | 1628 | |||
AK166557.1 | - | 5 | 1632 | |||
BC035295.1 | - | 5 | 1608 | 3985077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
