siRNA Details
Gene Symbol: Smagp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033872.2 | NP_001029044.1 | 4 | 401 | ||
NM_001285413.1 | NP_001272342.1 | 3 | 384 | |||
NM_001285414.1 | NP_001272343.1 | 3 | 337 | |||
NM_174992.4 | NP_778157.2 | 4 | 591 | |||
GenBank mRNA | BC004728.1 | - | 4 | 392 | 3582955 | |
BC132460.1 | - | 3 | 198 | 40130737 | ||
BC132462.1 | - | 3 | 198 | 40130739 | ||
BF018035.1 | - | 4 | 469 | |||
BY707988.1 | - | 4 | 326 | |||
CJ042753.1 | - | 4 | 354 | |||
CJ052870.1 | - | 3 | 279 | |||
CJ052873.1 | - | 3 | 385 | |||
CK129185.1 | - | 4 | 295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
