siRNA Details
Gene Symbol: 4930539E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172450.3 | NP_766038.1 | 2 | 610 | ||
GenBank mRNA | AK033585.1 | - | 2 | 611 | ||
BC117930.2 | - | 2 | 606 | 40097681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
