siRNA Details
Gene Symbol: Alg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243161.1 | NP_001230090.1 | 3 | 981 | ||
NM_183142.4 | NP_898965.1 | 3 | 855 | |||
GenBank mRNA | AB288237.1 | - | 3 | 975 | ||
AB288238.1 | - | 3 | 979 | |||
AK046475.1 | - | 3 | 857 | |||
AK134312.1 | - | 3 | 956 | |||
AK157746.1 | - | 3 | 981 | |||
BC061469.1 | - | 3 | 964 | 6821552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
