siRNA Details
Gene Symbol: Map6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198599.2 | NP_941001.2 | 1, 2 | 418 | ||
GenBank mRNA | AK079603.1 | - | 1, 2 | 381 | ||
AK163656.1 | - | 1, 2 | 382 | |||
BB653005.1 | - | 1, 2 | 419 | |||
BC116220.1 | - | 1, 2 | 381 | 40091320 | ||
BC116221.1 | - | 1, 2 | 381 | 40091321 | ||
BE979311.1 | - | 1, 2 | 92 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
