siRNA Details
Gene Symbol: Map6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198599.2 | NP_941001.2 | 3 | 1461 | ||
GenBank mRNA | AK163656.1 | - | 3 | 1425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
