siRNA Details
Gene Symbol: Gm609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005854.2 | NP_001005854.2 | 2, 3 | 455 | ||
GenBank mRNA | AB853320.1 | - | 2, 3 | 471 | ||
AK136536.1 | - | 2, 3 | 473 | |||
BC054854.1 | - | 2, 3 | 454 | 6529072 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
