siRNA Details
Gene Symbol: Gm609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005854.2 | NP_001005854.2 | 4 | 767 | ||
GenBank mRNA | AB853320.1 | - | 4 | 783 | ||
AK136536.1 | - | 4 | 785 | |||
BC054854.1 | - | 4 | 766 | 6529072 |
Gene Symbol: Gm17783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB853321.1 | - | 4 | 778 |
